Learn More
Sklar Instruments Econo Lister Bandage Scissors, Sklar, 5-1/2 in., Blunt/Blunt, Angled, Smooth, Finger Rings, Pocket Clip, Stainless Steel. PK/12.
SDP
Catalog No. 501437565 Shop All Sklar Instruments Products
Encompass_Preferred
Econo Lister Bandage Scissors, Sklar, 5-1/2 in., Blunt/Blunt, Angled, Smooth, Finger Rings, Pocket Clip, Stainless Steel. Econo Lister Bandage Scissors are a versatile instrument with multiple uses. They are commonly used for sizing dressings and removing circumferential bandages. They may also be used to cut tougher material such as plaster, fabric and occasionally umbilical cord. The jaws of the Lister Bandage Scissors are angled with the lower blade being slightly longer. The tip of the lower blade features a flattened blunt nodule that is intended to slide between bandages and skin without harming the sk in. This feature allows Lister Bandage Scissors to occasionally be used during cesarean section to open the uterus without causing inadvertent injury to the baby. Ring handles can be equal in size or with one ring larger. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. It features a pocket clip. They are sold in boxes of 12.
This item is not returnable. View return policy
Catalog No. 50-143-7565 Supplier Sklar Instruments Supplier No. 21237
Econo Lister Bandage Scissors, Sklar, 5-1/2 in., Blunt/Blunt, Angled, Smooth, Finger Rings, Pocket Clip, Stainless Steel. Econo Lister Bandage Scissors are a versatile instrument with multiple uses. They are commonly used for sizing dressings and removing circumferential bandages. They may also be used to cut tougher material such as plaster, fabric and occasionally umbilical cord. The jaws of the Lister Bandage Scissors are angled with the lower blade being slightly longer. The tip of the lower blade features a flattened blunt nodule that is intended to slide between bandages and skin without harming the sk in. This feature allows Lister Bandage Scissors to occasionally be used during cesarean section to open the uterus without causing inadvertent injury to the baby. Ring handles can be equal in size or with one ring larger. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. It features a pocket clip. They are sold in boxes of 12.