Learn More
Med Vet International Dynarex Sensi Wrap, Self-Adherent, 3" X 5 Yd Tan, 24/Case
Catalog No. 502829723
Encompass_Preferred
Dynarex Sensi-Wrap Self-Adherent Bandage Rolls are premium compression bandages designed to help treat a wide range of conditions including edema, general soreness, and swelling of joints. These high-quality Bandages help to securely hold splints, gel sheeting, primary dressings, or elastomer molds in place to help promote the healing process. These convenient self-wrapping bandages hold securely without the need for clips or fasteners. Dynarex Sensi-Wrap Self-Adherent Bandage Rolls tear easily without scissors for time-saving application. Dynarex Sensi-Wrap Bandages are available in a variety of colors.   Dynarex Sensi-Wrap Self-Adherent Bandage Rolls are lightweight bandages, used in both human and veterinary markets, designed to provide effective compression to help prevent and treat a variety of injuries in both the human and veterinary markets. These high-quality Bandages provide optimal pressure to help reduce swelling and prevent fluids from accumulating at the injury site.  
Catalog No. 50-282-9723 Supplier Med Vet International Supplier No. 3173
Dynarex Sensi-Wrap Self-Adherent Bandage Rolls are premium compression bandages designed to help treat a wide range of conditions including edema, general soreness, and swelling of joints. These high-quality Bandages help to securely hold splints, gel sheeting, primary dressings, or elastomer molds in place to help promote the healing process. These convenient self-wrapping bandages hold securely without the need for clips or fasteners. Dynarex Sensi-Wrap Self-Adherent Bandage Rolls tear easily without scissors for time-saving application. Dynarex Sensi-Wrap Bandages are available in a variety of colors.   Dynarex Sensi-Wrap Self-Adherent Bandage Rolls are lightweight bandages, used in both human and veterinary markets, designed to provide effective compression to help prevent and treat a variety of injuries in both the human and veterinary markets. These high-quality Bandages provide optimal pressure to help reduce swelling and prevent fluids from accumulating at the injury site.  