Learn More
Med Vet International Dynarex Sensi Wrap, Self-Adherent Wrap - Latex Free, 3" X 5 Yds Light Blue, 24/Case
Supplier:  Med Vet International 3318
Encompass_Preferred
Dynarex Sensi-Wrap Self-Adherent Bandage Rolls are premium compression bandages designed to help manage a wide range of conditions including edema, general soreness, and swelling of joints. These high-quality Bandages are constructed to securely hold splints, gel sheeting, primary dressings, or elastomer molds in place. The convenient self-wrapping bandages hold securely without the need for clips or fasteners. These Bandage Rolls tear easily without scissors for time-saving application. Dynarex Sensi-Wrap Bandages are available in multiple colors and kid-friendly designs  Dynarex Sensi-Wrap Self-Adherent Bandage Rolls are lightweight bandages designed to provide effective compression to help manage a variety of injuries. These high-quality Bandage are constructed to provide pressure to help reduce swelling and help resist fluids from accumulating at the injury site. Ideal for use in medical and veterinary environments. 
Catalog No. 50-283-1843