Learn More
Med Vet International Dynarex Bandage Shears, 5.5", Red, 50/Box
Catalog No. 502829267
Encompass_Preferred
Featuring razor-sharp hardened surgical stainless steel blade with one serrated edge. Razor sharp hardened surgical stainless steel blade with one serrated edge. Easily cuts gauze, tape and other small items. Strong enough to cut clothing and seat belts. Size: 7.5 inches and 5.5 inches Available in multiple colors; Wrapped in an individual poly bag. Can be sterilized by autoclaving.
Catalog No. 50-282-9267 Supplier Med Vet International Supplier No. 4196BX
Featuring razor-sharp hardened surgical stainless steel blade with one serrated edge. Razor sharp hardened surgical stainless steel blade with one serrated edge. Easily cuts gauze, tape and other small items. Strong enough to cut clothing and seat belts. Size: 7.5 inches and 5.5 inches Available in multiple colors; Wrapped in an individual poly bag. Can be sterilized by autoclaving.