Learn More
Med Vet International Dynarex Bandage Shears, 5.5" Teal, 200/Case
Catalog No. 502829335
Encompass_Preferred
Featuring razor-sharp hardened surgical stainless steel blade with one serrated edge. Razor sharp hardened surgical stainless steel blade with one serrated edge. Easily cuts gauze, tape and other small items. Strong enough to cut clothing and seat belts. Size: 7.5 inches and 5.5 inches Available in multiple colors; Wrapped in an individual poly bag. Can be sterilized by autoclaving.
Catalog No. 50-282-9335 Supplier Med Vet International Supplier No. 4197
Add to Cart
Add to Cart
Featuring razor-sharp hardened surgical stainless steel blade with one serrated edge. Razor sharp hardened surgical stainless steel blade with one serrated edge. Easily cuts gauze, tape and other small items. Strong enough to cut clothing and seat belts. Size: 7.5 inches and 5.5 inches Available in multiple colors; Wrapped in an individual poly bag. Can be sterilized by autoclaving.