Learn More
Med Vet International Unna'S Boot 3" X 10 Yds., Each
Catalog No. 502830712
Encompass_Preferred
Non-raveling gauze evenly impregnated with a non-hardening zinc oxide paste. Contains no preservatives. Without Calamine. 3" x 10 Yds., Each.
Catalog No. 50-283-0712 Supplier Med Vet International Supplier No. UNNA3
Non-raveling gauze evenly impregnated with a non-hardening zinc oxide paste. Contains no preservatives. Without Calamine. 3" x 10 Yds., Each.