Learn More
Med Vet International Miltex Vantage Universal Bandage and Utility Scissors, 7.5in, Blue Handle
Catalog No. 502522275
Encompass_Preferred
The Miltex Vantage V95-1027 universal bandage and utility scissors are 7.5" in length and have blue handles. They are designed to cut bandage and gauze, cast material, metal sheets, and tough clothing material. Autoclavable to 143C or 290F for easy sterilization.
Catalog No. 50-252-2275 Supplier Med Vet International Supplier No. MILV951027
The Miltex Vantage V95-1027 universal bandage and utility scissors are 7.5" in length and have blue handles. They are designed to cut bandage and gauze, cast material, metal sheets, and tough clothing material. Autoclavable to 143C or 290F for easy sterilization.