Learn More
Med Vet International Miltex Vantage Universal Bandage And Utility Scissors, 7.5in., Orange Handle
Catalog No. 502829055
Encompass_Preferred
The Miltex Vantage V95-1026 universal bandage and utility scissors are 7.5" in length and have orange handles. They are designed to cut bandage and gauze, cast material, metal sheets, and tough clothing material. Autoclavable to 143C or 290F for easy sterilization.
Catalog No. 50-282-9055 Supplier Med Vet International Supplier No. MILV951026
The Miltex Vantage V95-1026 universal bandage and utility scissors are 7.5" in length and have orange handles. They are designed to cut bandage and gauze, cast material, metal sheets, and tough clothing material. Autoclavable to 143C or 290F for easy sterilization.