Learn More
Med Vet International Waterproof Medical Tape Band-Aid Water Block™ White 1 Inch X 10 Yard Adhesive NonSterile, 38137117121, Each
Catalog No. 503044025
Encompass_Preferred
Adheres gently and securely to intact skin Easy to tear to the right size Won't stick to wound bed Absolute waterproof tape secures gauze in place for burns or cuts The hypoallergenic medical tape is latex-free, making it gentle on the skin The tape is flexible to fit in any part of the body It's breathable to increase the wearer's comfort The strong adhesive on the tape ensures it stays on during activities 1 inch x 30 feet
Catalog No. 50-304-4025 Supplier Med Vet International Supplier No. 1169364EA
Adheres gently and securely to intact skin Easy to tear to the right size Won't stick to wound bed Absolute waterproof tape secures gauze in place for burns or cuts The hypoallergenic medical tape is latex-free, making it gentle on the skin The tape is flexible to fit in any part of the body It's breathable to increase the wearer's comfort The strong adhesive on the tape ensures it stays on during activities 1 inch x 30 feet