Learn More
Grainger Dayton Banded V-Belt, C Section, 2 Ribs, 127.9 in. Length, 1 ea
Catalog No. 502770860
Encompass_Preferred
This banded V-belt features a classical C-section design, engineered by fusing two single V-belts together to provide stability and power transmission in multi-belt systems. Constructed from a durable rubber compound with polyester reinforcing cords, it is designed for use in industrial applications such as conveyors, blowers, and heavy machinery.
  • Features a 1-3/4 in. top width and 5/8 in. thickness
  • Includes 2 ribs for enhanced power distribution
  • Provides an outside length of 127.9 in.
  • Resists heat, oil, and abrasion for reliable operation
  • Operates within a temperature range of -22°F to 194°F
  • Utilizes a reinforced tie band and wrapped edge construction
Catalog No. 50-277-0860 Supplier Grainger Supplier No. 13W390
This banded V-belt features a classical C-section design, engineered by fusing two single V-belts together to provide stability and power transmission in multi-belt systems. Constructed from a durable rubber compound with polyester reinforcing cords, it is designed for use in industrial applications such as conveyors, blowers, and heavy machinery.
  • Features a 1-3/4 in. top width and 5/8 in. thickness
  • Includes 2 ribs for enhanced power distribution
  • Provides an outside length of 127.9 in.
  • Resists heat, oil, and abrasion for reliable operation
  • Operates within a temperature range of -22°F to 194°F
  • Utilizes a reinforced tie band and wrapped edge construction