Learn More
Grainger Dayton Banded V-Belt, C Section, 2 Ribs, 272 in Length, 1 3/4 in Top Width, 1 ea
Catalog No. 502773210
Encompass_Preferred
These C section banded V-belts are designed for medium-horsepower drive systems in applications such as conveyors, industrial blowers, and machinery. The construction consists of multiple V-belts fused together with a reinforced backing to provide stability and power transmission efficiency.
  • Features 2 ribs for reliable performance in multiple belt systems
  • Rubber compound construction resists abrasion, oils, coolants, and ozone exposure
  • Polyester reinforcing cord offers high heat- and stretch-resistance
  • Operates in temperatures ranging from -22°F to 194°F
  • Wrapped edge construction for durability
Catalog No. 50-277-3210 Supplier Grainger Supplier No. 13W407
These C section banded V-belts are designed for medium-horsepower drive systems in applications such as conveyors, industrial blowers, and machinery. The construction consists of multiple V-belts fused together with a reinforced backing to provide stability and power transmission efficiency.
  • Features 2 ribs for reliable performance in multiple belt systems
  • Rubber compound construction resists abrasion, oils, coolants, and ozone exposure
  • Polyester reinforcing cord offers high heat- and stretch-resistance
  • Operates in temperatures ranging from -22°F to 194°F
  • Wrapped edge construction for durability