Learn More
Grainger Dayton Banded V-Belt, C Section, 2 Ribs, 332.9 in. Outside Length, 1 ea.
Catalog No. 502770865
Encompass_Preferred
This banded V-belt features a classical C-section design, constructed by fusing two individual V-belts together to create a robust, integrated component. Ideal for medium-horsepower drive systems, this belt provides reliable performance in industrial applications such as conveyors, blowers, and heavy machinery.
  • Features a 2-rib configuration with a reinforced tie band for durability
  • Constructed from high-quality rubber compound for resistance against heat, oil, and ozone exposure
  • Utilizes polyester reinforcing cord for improved stretch and heat resistance
  • Operates within a temperature range of -22°F to +194°F
  • Designed with a 332.9 in. outside length and 1-3/4 in. top width
  • Wrapped edge construction for consistent grip in power transmission systems
Catalog No. 50-277-0865 Supplier Grainger Supplier No. 13W411
This banded V-belt features a classical C-section design, constructed by fusing two individual V-belts together to create a robust, integrated component. Ideal for medium-horsepower drive systems, this belt provides reliable performance in industrial applications such as conveyors, blowers, and heavy machinery.
  • Features a 2-rib configuration with a reinforced tie band for durability
  • Constructed from high-quality rubber compound for resistance against heat, oil, and ozone exposure
  • Utilizes polyester reinforcing cord for improved stretch and heat resistance
  • Operates within a temperature range of -22°F to +194°F
  • Designed with a 332.9 in. outside length and 1-3/4 in. top width
  • Wrapped edge construction for consistent grip in power transmission systems