Learn More
Grainger Dayton Banded V-Belt, 3 Ribs, 61.8 in Outside Length, 2 in Top Width, 1 ea
Catalog No. 502772143
Encompass_Preferred
This B section banded V-belt features a classical design composed of three single V-belts fused together to form a robust, three-rib assembly with reinforced backing. Engineered for medium-horsepower drive systems, it is suitable for industrial applications including blowers, conveyors, and various machinery. The belt is constructed from a high-performance rubber compound reinforced with polyester cords to ensure durability and reliability under load.
  • Banded design with 3 ribs for stable power transmission.
  • Rubber compound provides resistance to abrasion, oils, coolants, ozone exposure, and heat.
  • Polyester reinforcing cords offer stretch-resistance and strength.
  • Temperature operating range of -22°F to +194°F.
  • Wrapped edge construction with a reinforced tie band.
Catalog No. 50-277-2143 Supplier Grainger Supplier No. 13V476
This B section banded V-belt features a classical design composed of three single V-belts fused together to form a robust, three-rib assembly with reinforced backing. Engineered for medium-horsepower drive systems, it is suitable for industrial applications including blowers, conveyors, and various machinery. The belt is constructed from a high-performance rubber compound reinforced with polyester cords to ensure durability and reliability under load.
  • Banded design with 3 ribs for stable power transmission.
  • Rubber compound provides resistance to abrasion, oils, coolants, ozone exposure, and heat.
  • Polyester reinforcing cords offer stretch-resistance and strength.
  • Temperature operating range of -22°F to +194°F.
  • Wrapped edge construction with a reinforced tie band.