Learn More
Grainger Dayton Banded V-Belt, B Section, 3 Ribs, 62.8 in. Outside Length, 1 ea
Catalog No. 502772552
Encompass_Preferred
This B section banded V-belt features a classical design consisting of three individual V-belts fused together to form a cohesive unit with a reinforced tie band. Engineered for medium-horsepower drive systems, this belt provides consistent power transmission in demanding industrial applications such as blowers, conveyors, and various machinery.
  • Features 3 ribs with a 2 in. top width and 62.8 in. outside length.
  • Constructed from durable rubber compound for high heat tolerance and resistance to abrasion, oils, and coolants.
  • Includes polyester reinforcing cords for enhanced heat and stretch resistance.
  • Designed for operation in temperatures ranging from -22°F to +194°F.
  • Features a 1/2 in. thickness and a wrapped edge construction for reliable performance.
Catalog No. 50-277-2552 Supplier Grainger Supplier No. 13V477
This B section banded V-belt features a classical design consisting of three individual V-belts fused together to form a cohesive unit with a reinforced tie band. Engineered for medium-horsepower drive systems, this belt provides consistent power transmission in demanding industrial applications such as blowers, conveyors, and various machinery.
  • Features 3 ribs with a 2 in. top width and 62.8 in. outside length.
  • Constructed from durable rubber compound for high heat tolerance and resistance to abrasion, oils, and coolants.
  • Includes polyester reinforcing cords for enhanced heat and stretch resistance.
  • Designed for operation in temperatures ranging from -22°F to +194°F.
  • Features a 1/2 in. thickness and a wrapped edge construction for reliable performance.