Learn More
Grainger Dayton Banded V-Belt, B Section, 3 Ribs, 68.8 in Outside Length, 1 ea
Catalog No. 502771056
Encompass_Preferred
This B-section banded V-belt features a classical design, utilizing three individual V-belts fused together to form a cohesive, ribbed belt. It is engineered for medium-horsepower drive systems commonly found in industrial machinery, blowers, and conveyor applications. The belt incorporates a rubber compound body designed to provide durability and resistance against heat, abrasion, oil, and coolant, while polyester cords offer reliable strength and stretch-resistance during operation.
  • Banded design with 3 ribs for stable power transmission
  • Constructed with rubber compound and polyester reinforcing cords
  • Operates in temperatures from -22°F to +194°F
  • Wrapped edge construction with reinforced tie band
  • Outside length of 68.8 in. and nominal length of 68 in.
  • Top width of 2 in. and thickness of 0.5 in.
Catalog No. 50-277-1056 Supplier Grainger Supplier No. 13V481
This B-section banded V-belt features a classical design, utilizing three individual V-belts fused together to form a cohesive, ribbed belt. It is engineered for medium-horsepower drive systems commonly found in industrial machinery, blowers, and conveyor applications. The belt incorporates a rubber compound body designed to provide durability and resistance against heat, abrasion, oil, and coolant, while polyester cords offer reliable strength and stretch-resistance during operation.
  • Banded design with 3 ribs for stable power transmission
  • Constructed with rubber compound and polyester reinforcing cords
  • Operates in temperatures from -22°F to +194°F
  • Wrapped edge construction with reinforced tie band
  • Outside length of 68.8 in. and nominal length of 68 in.
  • Top width of 2 in. and thickness of 0.5 in.