Learn More
Grainger Dayton Banded V-Belt, 3 Ribs, C Section, 377.9 in. Length, 1 ea
Catalog No. 502770742
Encompass_Preferred
This banded C-section V-belt features a classical design consisting of three individual belts fused together with a reinforced backing to create a stable, multi-ribbed system. Ideal for medium-horsepower drive systems, these belts are engineered for consistent performance in industrial machinery, conveyors, and blowers. The durable rubber compound construction resists abrasion, oils, coolants, and ozone, while polyester reinforcing cords provide necessary heat and stretch resistance.
  • Features 3 ribs for reliable power transmission
  • Includes reinforced tie band construction for durability
  • Operates in temperatures ranging from -22°F to +194°F
  • Utilizes oil- and heat-resistant rubber materials
  • Designed for use in multiple belt systems
Catalog No. 50-277-0742 Supplier Grainger Supplier No. 13W448
This banded C-section V-belt features a classical design consisting of three individual belts fused together with a reinforced backing to create a stable, multi-ribbed system. Ideal for medium-horsepower drive systems, these belts are engineered for consistent performance in industrial machinery, conveyors, and blowers. The durable rubber compound construction resists abrasion, oils, coolants, and ozone, while polyester reinforcing cords provide necessary heat and stretch resistance.
  • Features 3 ribs for reliable power transmission
  • Includes reinforced tie band construction for durability
  • Operates in temperatures ranging from -22°F to +194°F
  • Utilizes oil- and heat-resistant rubber materials
  • Designed for use in multiple belt systems