Learn More
Grainger Dayton Banded V-Belt, 8V Section, 4 Ribs, 171 in. Length, 1 ea
Catalog No. 502773024
Encompass_Preferred
This banded V-belt features a wedge design composed of four individual 8V section V-belts fused together to create a robust, four-rib assembly. Engineered for high power transmission, it offers a deeper profile and greater efficiency than classical V-belt designs. The belt is constructed from a durable rubber compound reinforced with heat- and stretch-resistant polyester cords.
  • Banded design with 4 ribs for superior power transfer
  • Constructed from high-tolerance rubber compound and polyester reinforcing cords
  • Resistant to heat, oil, abrasion, coolants, and ozone
  • Wrapped edge construction with bias cut cover fabric
  • Operating temperature range of -22°F to +194°F
  • Outside length of 171 in. with 4 in. top width
Catalog No. 50-277-3024 Supplier Grainger Supplier No. 13V619
This banded V-belt features a wedge design composed of four individual 8V section V-belts fused together to create a robust, four-rib assembly. Engineered for high power transmission, it offers a deeper profile and greater efficiency than classical V-belt designs. The belt is constructed from a durable rubber compound reinforced with heat- and stretch-resistant polyester cords.
  • Banded design with 4 ribs for superior power transfer
  • Constructed from high-tolerance rubber compound and polyester reinforcing cords
  • Resistant to heat, oil, abrasion, coolants, and ozone
  • Wrapped edge construction with bias cut cover fabric
  • Operating temperature range of -22°F to +194°F
  • Outside length of 171 in. with 4 in. top width