Learn More
Grainger Dayton Banded V-Belt, 8V Section, 4 Ribs, 181 in Length, 1 ea
Catalog No. 502771109
Encompass_Preferred
This banded wedge V-belt is engineered for heavy-duty industrial applications requiring high power transmission and vibration resistance. Constructed with a robust rubber body reinforced by polyester cords, it offers durability and efficient performance in demanding drive systems.
  • Banded design with 4 ribs for increased stability and uniform load distribution
  • Constructed from rubber and polyester cords for reliable strength
  • Features heat and oil resistance for longevity in harsh environments
  • Wrapped edge construction ensures consistent grip and extended service life
  • Operating temperature range of -22 °F to +194 °F
  • Outside length of 181 in., top width of 4 in., and thickness of 1 in.
Catalog No. 50-277-1109 Supplier Grainger Supplier No. 13V620
This banded wedge V-belt is engineered for heavy-duty industrial applications requiring high power transmission and vibration resistance. Constructed with a robust rubber body reinforced by polyester cords, it offers durability and efficient performance in demanding drive systems.
  • Banded design with 4 ribs for increased stability and uniform load distribution
  • Constructed from rubber and polyester cords for reliable strength
  • Features heat and oil resistance for longevity in harsh environments
  • Wrapped edge construction ensures consistent grip and extended service life
  • Operating temperature range of -22 °F to +194 °F
  • Outside length of 181 in., top width of 4 in., and thickness of 1 in.