Learn More
Grainger Dayton Banded V-Belt, B Section, 4 Ribs, 80.8 in. Length, 1 ea
Catalog No. 502770982
Encompass_Preferred
This B section banded V-belt features a classical design constructed of 4 individual belts fused with a reinforced backing to ensure synchronized performance in medium-horsepower drive systems. Designed for use in industrial machinery, blowers, and conveyor applications, this belt provides reliable power transmission through a 4-rib configuration. It is built to endure demanding environments with resistance to heat, oil, coolants, and ozone.
  • 4-rib design for consistent belt tracking and load distribution
  • Rubber compound construction resists flex fatigue and abrasion
  • Polyester reinforcing cords provide high strength and stretch resistance
  • Operating temperature range of -22°F to +194°F
  • Wrapped edge construction for enhanced durability
Catalog No. 50-277-0982 Supplier Grainger Supplier No. 13V548
This B section banded V-belt features a classical design constructed of 4 individual belts fused with a reinforced backing to ensure synchronized performance in medium-horsepower drive systems. Designed for use in industrial machinery, blowers, and conveyor applications, this belt provides reliable power transmission through a 4-rib configuration. It is built to endure demanding environments with resistance to heat, oil, coolants, and ozone.
  • 4-rib design for consistent belt tracking and load distribution
  • Rubber compound construction resists flex fatigue and abrasion
  • Polyester reinforcing cords provide high strength and stretch resistance
  • Operating temperature range of -22°F to +194°F
  • Wrapped edge construction for enhanced durability