Learn More
Grainger Dayton Banded V-Belt, Rubber Compound, 98.8 in. Length, 4 Ribs
Catalog No. 502770596
Encompass_Preferred
This banded V-belt is designed for reliable power transmission in multiple belt systems, offering increased stability through its four-ribbed construction. It features a durable rubber compound body with polyester reinforcing cords to ensure strength and longevity in demanding industrial applications.
  • Banded V-belt design with 4 ribs
  • Constructed from durable rubber compound with polyester cords
  • Features wrapped edge construction for added protection
  • Oil-resistant and heat-resistant properties
  • Operating temperature range of -22°F to +194°F
  • Industry number 4/B95
  • Outside length of 98.8 in.
  • Thickness of 1/2 in. and top width of 2 43/64 in.
Catalog No. 50-277-0596 Supplier Grainger Supplier No. 13V558
This banded V-belt is designed for reliable power transmission in multiple belt systems, offering increased stability through its four-ribbed construction. It features a durable rubber compound body with polyester reinforcing cords to ensure strength and longevity in demanding industrial applications.
  • Banded V-belt design with 4 ribs
  • Constructed from durable rubber compound with polyester cords
  • Features wrapped edge construction for added protection
  • Oil-resistant and heat-resistant properties
  • Operating temperature range of -22°F to +194°F
  • Industry number 4/B95
  • Outside length of 98.8 in.
  • Thickness of 1/2 in. and top width of 2 43/64 in.