Learn More
Grainger Dayton Banded V-Belt, 4 Ribs, 100.8 in. Length, 1/2 in. Thickness, 1 ea
Catalog No. 502770654
Encompass_Preferred
This banded V-belt is designed for use in multiple belt systems, providing reliable power transmission in demanding industrial applications. The belt features a durable rubber compound construction with a wrapped edge and reinforcing polyester cords for added strength and stability. It is engineered to perform in diverse environments, offering resistance to both heat and oil.
  • Banded design with 4 ribs
  • Fabric cover with rubber body and polyester cords
  • Heat-resistant and oil-resistant properties
  • Operating temperature range of -22°F to +194°F
  • Industry number 4/B97
  • Outside length of 100.8 in.
  • Top width of 2 43/64 in. and thickness of 1/2 in.
Catalog No. 50-277-0654 Supplier Grainger Supplier No. 13V560
This banded V-belt is designed for use in multiple belt systems, providing reliable power transmission in demanding industrial applications. The belt features a durable rubber compound construction with a wrapped edge and reinforcing polyester cords for added strength and stability. It is engineered to perform in diverse environments, offering resistance to both heat and oil.
  • Banded design with 4 ribs
  • Fabric cover with rubber body and polyester cords
  • Heat-resistant and oil-resistant properties
  • Operating temperature range of -22°F to +194°F
  • Industry number 4/B97
  • Outside length of 100.8 in.
  • Top width of 2 43/64 in. and thickness of 1/2 in.