Learn More
Grainger Dayton Banded V-Belt, C Section, 4 Ribs, 109.9 in. Length, 1 ea
Catalog No. 502771773
Encompass_Preferred
This banded V-belt is designed for efficient power transmission in multiple belt systems, providing reliable performance and consistent tension. Constructed with high-quality materials, it offers durability and resistance to challenging operating environments.
  • Features a reinforced tie band construction for improved stability and alignment.
  • Equipped with 4 ribs for effective power distribution.
  • Constructed from rubber impregnated fabric with polyester cords for enhanced strength.
  • Offers resistance to oil and heat.
  • Operating temperature range from -22°F to +194°F.
  • Wrapped edge construction for wear resistance.
Catalog No. 50-277-1773 Supplier Grainger Supplier No. 13W454
This banded V-belt is designed for efficient power transmission in multiple belt systems, providing reliable performance and consistent tension. Constructed with high-quality materials, it offers durability and resistance to challenging operating environments.
  • Features a reinforced tie band construction for improved stability and alignment.
  • Equipped with 4 ribs for effective power distribution.
  • Constructed from rubber impregnated fabric with polyester cords for enhanced strength.
  • Offers resistance to oil and heat.
  • Operating temperature range from -22°F to +194°F.
  • Wrapped edge construction for wear resistance.