Learn More
Grainger Dayton Banded V-Belt, 4 Ribs, C Section, 128.9 in Length, 1 ea
Catalog No. 502770069
Encompass_Preferred
This banded V-belt features a classical C-section design, constructed by fusing four individual V-belts together to provide enhanced stability for medium-horsepower drive systems. It is ideal for use in industrial applications such as conveyors, blowers, and machinery where reliable power transmission is required. The durable construction resists abrasion, oils, and coolants while offering high heat tolerance for demanding environments.
  • 4 ribs for uniform power distribution
  • 128.9 in. outside length
  • 3-1/2 in. top width and 5/8 in. thickness
  • Reinforced with polyester cords for stretch-resistance
  • Temperature range of -22°F to +194°F
  • Heat- and oil-resistant rubber compound construction
  • Wrapped edge construction for increased durability
Catalog No. 50-277-0069 Supplier Grainger Supplier No. 13W460
This banded V-belt features a classical C-section design, constructed by fusing four individual V-belts together to provide enhanced stability for medium-horsepower drive systems. It is ideal for use in industrial applications such as conveyors, blowers, and machinery where reliable power transmission is required. The durable construction resists abrasion, oils, and coolants while offering high heat tolerance for demanding environments.
  • 4 ribs for uniform power distribution
  • 128.9 in. outside length
  • 3-1/2 in. top width and 5/8 in. thickness
  • Reinforced with polyester cords for stretch-resistance
  • Temperature range of -22°F to +194°F
  • Heat- and oil-resistant rubber compound construction
  • Wrapped edge construction for increased durability