Learn More
Grainger Dayton Banded V-Belt, 8V Section, 5 Ribs, 561 in. Outside Length, 1 ea
Catalog No. 502773401
Encompass_Preferred
The banded V-belt is engineered for high-power transfer in multiple belt systems, featuring a wedge design that enhances performance over classical V-belt profiles. Constructed with five V-belts fused into a single unit, this component provides stability and efficient power transmission. It utilizes a durable rubber compound capable of withstanding significant flex fatigue and elevated heat, while offering resistance to abrasion, oils, coolants, and ozone exposure.
  • Features 5 ribs with a 561 in. outside length and 5 in. top width
  • Reinforced with polyester cord for heat and stretch resistance
  • Operates in a temperature range of -22 °F to 194 °F
  • Wrapped edge construction ensures durability in demanding applications
  • Designed for use with multiple belt systems
Catalog No. 50-277-3401 Supplier Grainger Supplier No. 13V670
Add to Cart
Add to Cart
Request a Quote
The banded V-belt is engineered for high-power transfer in multiple belt systems, featuring a wedge design that enhances performance over classical V-belt profiles. Constructed with five V-belts fused into a single unit, this component provides stability and efficient power transmission. It utilizes a durable rubber compound capable of withstanding significant flex fatigue and elevated heat, while offering resistance to abrasion, oils, coolants, and ozone exposure.
  • Features 5 ribs with a 561 in. outside length and 5 in. top width
  • Reinforced with polyester cord for heat and stretch resistance
  • Operates in a temperature range of -22 °F to 194 °F
  • Wrapped edge construction ensures durability in demanding applications
  • Designed for use with multiple belt systems