Learn More
Grainger Dayton Banded V-Belt, B Section, 5 Ribs, 49.8 in. Outside Length, 1 ea.
Catalog No. 502773509
Encompass_Preferred
This banded B-section V-belt features a classical design constructed of multiple V-belts fused together to form a unit with five ribs and a reinforced backing. It is designed for use in medium-horsepower drive systems such as conveyors, industrial blowers, and machinery.
  • Features five ribs with a 3 21/64 in. top width and 1/2 in. thickness.
  • Constructed from rubber compound that withstands flex fatigue and provides high heat tolerance.
  • Resistant to abrasion, oils, coolants, and ozone exposure.
  • Reinforced with polyester cord for heat- and stretch-resistance.
  • Operating temperature range of -22 °F to +194 °F.
  • Designed for 49.8 in. outside length.
Catalog No. 50-277-3509 Supplier Grainger Supplier No. 13V580
This banded B-section V-belt features a classical design constructed of multiple V-belts fused together to form a unit with five ribs and a reinforced backing. It is designed for use in medium-horsepower drive systems such as conveyors, industrial blowers, and machinery.
  • Features five ribs with a 3 21/64 in. top width and 1/2 in. thickness.
  • Constructed from rubber compound that withstands flex fatigue and provides high heat tolerance.
  • Resistant to abrasion, oils, coolants, and ozone exposure.
  • Reinforced with polyester cord for heat- and stretch-resistance.
  • Operating temperature range of -22 °F to +194 °F.
  • Designed for 49.8 in. outside length.