Learn More
Grainger Grainger Inc. Banded V-Belt, 5-Rib, 232 in., 1/Pk.
Catalog No. 502773442
Encompass_Preferred
This banded V-belt is designed for reliable power transmission in demanding industrial applications. The multi-belt construction provides stability and consistent performance across heavy-duty drive systems.
  • Banded construction for reduced vibration and improved stability
  • Designed for 5/C profile drive systems
  • Effective length of 232 in.
  • Engineered for high-torque industrial equipment
Catalog No. 50-277-3442 Supplier Grainger Supplier No. 13W507
This banded V-belt is designed for reliable power transmission in demanding industrial applications. The multi-belt construction provides stability and consistent performance across heavy-duty drive systems.
  • Banded construction for reduced vibration and improved stability
  • Designed for 5/C profile drive systems
  • Effective length of 232 in.
  • Engineered for high-torque industrial equipment