Learn More
Grainger Dayton Banded V-Belt, B Section, 6 Ribs, 106.8 in. Outside Length
Catalog No. 502773658
Encompass_Preferred
This banded V-belt is designed for reliable power transmission in industrial applications. Featuring a reinforced tie-band construction, it ensures stable performance under demanding conditions.
  • Banded V-belt type with 6 ribs
  • Belt section B
  • Industry number 6/B103
  • Constructed from rubber impregnated fabric with polyester cords
  • Wrapped edge construction for durability
  • Heat and oil resistant properties
  • Operating temperature range of -22°F to +194°F
  • Outside length of 106 4/5 in.
  • Thickness of 1/2 in. and top width of 4 in.
Catalog No. 50-277-3658 Supplier Grainger Supplier No. 13W359
This banded V-belt is designed for reliable power transmission in industrial applications. Featuring a reinforced tie-band construction, it ensures stable performance under demanding conditions.
  • Banded V-belt type with 6 ribs
  • Belt section B
  • Industry number 6/B103
  • Constructed from rubber impregnated fabric with polyester cords
  • Wrapped edge construction for durability
  • Heat and oil resistant properties
  • Operating temperature range of -22°F to +194°F
  • Outside length of 106 4/5 in.
  • Thickness of 1/2 in. and top width of 4 in.