Learn More
Grainger Dayton Banded V-Belt, B Section, 6 Ribs, 161 in. Length, 1 ea
Catalog No. 502770182
Encompass_Preferred
This banded V-belt features a classical B-section design, constructed by fusing six individual V-belts together to form a cohesive unit with reinforced backing. Engineered for medium-horsepower drive systems, it is suitable for use in industrial blowers, conveyors, and various machinery. The rubber compound construction provides durability against flex fatigue and harsh environmental factors, while the polyester cord reinforcement offers reliable stretch and heat resistance.
  • Banded construction with 6 ribs for stable power transmission
  • Resistant to heat, oil, abrasions, ozone, and coolants
  • Operates efficiently in temperatures from -22°F to 194°F
  • Designed for B-section applications with a 161 in. outside length
Catalog No. 50-277-0182 Supplier Grainger Supplier No. 13W374
This banded V-belt features a classical B-section design, constructed by fusing six individual V-belts together to form a cohesive unit with reinforced backing. Engineered for medium-horsepower drive systems, it is suitable for use in industrial blowers, conveyors, and various machinery. The rubber compound construction provides durability against flex fatigue and harsh environmental factors, while the polyester cord reinforcement offers reliable stretch and heat resistance.
  • Banded construction with 6 ribs for stable power transmission
  • Resistant to heat, oil, abrasions, ozone, and coolants
  • Operates efficiently in temperatures from -22°F to 194°F
  • Designed for B-section applications with a 161 in. outside length