Learn More
Grainger Dayton Banded V-Belt, B Section, 6 Ribs, 90.8 in. Outside Length
Catalog No. 502770543
Encompass_Preferred
This banded V-belt is designed for medium-horsepower drive systems in applications such as conveyors, industrial blowers, and machinery. It features a classical B-section construction consisting of six V-belts fused together to form a single unit with a reinforced tie band for stability. The belt is engineered for performance in demanding conditions with a focus on durability and heat resistance.
  • 6-rib construction for consistent performance in multiple belt systems
  • Rubber compound material resists flex fatigue, abrasion, oils, coolants, and ozone exposure
  • Polyester reinforcing cord provides strength and stretch resistance
  • Operates in a temperature range of -22°F to +194°F
  • Wrapped edge construction with 90 in. nominal length and 90.8 in. outside length
Catalog No. 50-277-0543 Supplier Grainger Supplier No. 13W351
This banded V-belt is designed for medium-horsepower drive systems in applications such as conveyors, industrial blowers, and machinery. It features a classical B-section construction consisting of six V-belts fused together to form a single unit with a reinforced tie band for stability. The belt is engineered for performance in demanding conditions with a focus on durability and heat resistance.
  • 6-rib construction for consistent performance in multiple belt systems
  • Rubber compound material resists flex fatigue, abrasion, oils, coolants, and ozone exposure
  • Polyester reinforcing cord provides strength and stretch resistance
  • Operates in a temperature range of -22°F to +194°F
  • Wrapped edge construction with 90 in. nominal length and 90.8 in. outside length