Learn More
Med Vet International Elastic Bandage 3M Ace 3 inch Width Clip Detached Closure Tan Nonsterile Standard Compression, 207314, Each
Catalog No. 502829886
Encompass_Preferred
Innovative clip snaps in place, holds securely and is easily adjusted Provides moderate support to weak, sore muscles and joints Comfortable design is intended for prolonged wear Easy-to-use wrap design Fits discreetly under clothing Wrap conforms to multiple body parts for versatile relief Soft feel is comfortable against your skin Wash and reuse again and again 3 inches x 5-1/3 feet
Catalog No. 50-282-9886 Supplier Med Vet International Supplier No. 1084230EA
Innovative clip snaps in place, holds securely and is easily adjusted Provides moderate support to weak, sore muscles and joints Comfortable design is intended for prolonged wear Easy-to-use wrap design Fits discreetly under clothing Wrap conforms to multiple body parts for versatile relief Soft feel is comfortable against your skin Wash and reuse again and again 3 inches x 5-1/3 feet