Learn More
Grainger Banding Clip 5/8 PK100
Catalog No. 502765948
Encompass_Preferred
The Band-It Banding Clip fits 5/8" strap and is made of stainless steel. It has a width of 1-1/4", a height of 1-1/8", and a weight of 0.06 lbs. This banding clip is used to connect and tension banding straps. It can withstand up to 350 lbs of tension. The clip has rounded edges for comfort and safety. It is manufactured in the USA by Band-It. The clip is sold individually.
Catalog No. 50-276-5948 Supplier Grainger Supplier No. 16P347
The Band-It Banding Clip fits 5/8" strap and is made of stainless steel. It has a width of 1-1/4", a height of 1-1/8", and a weight of 0.06 lbs. This banding clip is used to connect and tension banding straps. It can withstand up to 350 lbs of tension. The clip has rounded edges for comfort and safety. It is manufactured in the USA by Band-It. The clip is sold individually.