Learn More
Stoelting™ Bane Bone Rongeurs
SDP
Catalog No. 10001974 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Curved
Catalog No. Model
10-001-974 Curved
1 options
Catalog No. 10-001-974 Supplier Stoelting™ Supplier No. 52165P
Add to Cart
Add to Cart
Rongeurs are strongly constructed instruments with a sharp-edged, scoop-shaped tip, used for gouging out bone.
  • Used to open a window in bone, often in the skull
  • Used in neurosurgery and orthopedic surgery to expose areas for operation or to aid in removal of the brain.
Specifications
Size 7 in.
Model Curved