Learn More
Aviva Systems Biology Corp Banf2 Antibody - N-terminal region (ARP58592_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58592_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Banf2. It was validated on Western Blot. . Alternate names: 4930517K23Rik, Baf-L, Baf-like, Gm115, RP23-17O9.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-488