Learn More
New England Biolabs, Inc. BanI – 5000 units
SDP
Catalog No. 50811957
Encompass
A restriction endonuclease that recognizes the sequence G^GYRC_C.
Catalog No. 50-811-957 Supplier New England Biolabs, Inc. Supplier No. R0118S
A restriction endonuclease that recognizes the sequence G^GYRC_C.