Learn More
New England Biolabs, Inc. BanII – 2000 units
SDP
Catalog No. 50811959
Encompass
A restriction endonuclease that recognizes the sequence G_RGCY^C.
Catalog No. 50-811-959 Supplier New England Biolabs, Inc. Supplier No. R0119S
A restriction endonuclease that recognizes the sequence G_RGCY^C.