Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Banoxantrone-d12 dihydrochloride | 1562066-98-1 | 95.0% | C22H18D12Cl2N4O6 | 10MG
SDP

Catalog No. 5000259971
Encompass_Preferred

Banoxantrone-d12 dihydrochloride is the deuterium-labeled form of the bioreductive prodrug banoxantrone (AQ4N), supplied as a solid dihydrochloride for research use. It is intended for isotope tracing, mass-shift experiments, metabolic stability, and pharmacokinetic studies, and is provided with ~95% purity (CAS 1562066-98-1).

  • Deuterium-labeled for isotope tracing and mass-shift experiments.
  • Suitable for metabolic stability and pharmacokinetic studies.
  • High purity for reliable analytical and mass spectrometry results.
  • Solid dihydrochloride form for convenient handling and storage.
  • Well characterized with molecular formula C22H18D12Cl2N4O6 for unambiguous identification.

Catalog No. 50-002-59971 Supplier Medchemexpress LLC Supplier No. HY13562AS10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Banoxantrone-d12 dihydrochloride is the deuterium-labeled form of the bioreductive prodrug banoxantrone (AQ4N), supplied as a solid dihydrochloride for research use. It is intended for isotope tracing, mass-shift experiments, metabolic stability, and pharmacokinetic studies, and is provided with ~95% purity (CAS 1562066-98-1).

  • Deuterium-labeled for isotope tracing and mass-shift experiments.
  • Suitable for metabolic stability and pharmacokinetic studies.
  • High purity for reliable analytical and mass spectrometry results.
  • Solid dihydrochloride form for convenient handling and storage.
  • Well characterized with molecular formula C22H18D12Cl2N4O6 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.