Learn More
Abcam Bano x antrone dihydrochloride (AQ4N), Topoisomerase II inhibitor, 50MG
Catalog No. 502705612
Encompass_Preferred
MW 517.4 Da, Purity >97%. Bioreductive AQ4 precursor. Topoisomerase II inhibitor. Converts under hypoxia to DNA-affinic mitoxantrone (ab141041) structural analog. Induces DNA intercalation. Shows antitumor effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5612 Supplier Abcam Supplier No. AB14457350MG
MW 517.4 Da, Purity >97%. Bioreductive AQ4 precursor. Topoisomerase II inhibitor. Converts under hypoxia to DNA-affinic mitoxantrone (ab141041) structural analog. Induces DNA intercalation. Shows antitumor effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement