Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bansyl Chloride 98.0+%, TCI America™
SDP

Catalog No. B09945G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09945G 5 g
B09941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B09945G Supplier TCI America Supplier No. B09945G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 43040-76-2
Molecular Formula C18H24ClNO2S
Molecular Weight (g/mol) 353.91
MDL Number MFCD00059134
InChI Key PIZHBGYIKYGXBV-UHFFFAOYSA-N
Synonym 5-(Dibutylamino)naphthalene-1-sulfonyl Chloride
PubChem CID 191102
IUPAC Name 5-(dibutylamino)naphthalene-1-sulfonyl chloride
SMILES CCCCN(CCCC)C1=C2C=CC=C(C2=CC=C1)S(Cl)(=O)=O

Specifications

Quantity 5 g
UN Number 3265
Formula Weight 353.91
Percent Purity ≥98.0% (HPLC)
Physical Form Liquid
Chemical Name or Material Bansyl Chloride [N-Protecting Agent for Peptide Research]
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.