Learn More
Medchemexpress LLC HY-N0011 10mg Medchemexpress, Baohuoside I CAS:113558-15-9 Purity:>98%
SDP
Catalog No. 501933352
Encompass_Preferred
Medchemexpress, HY-N0011 10mg Baohuoside I CAS:113558-15-9 Baohuoside I, a flavonoid isolated from Epimedium koreanum Nakai, acts as an inhibitor of CXCR4 , downregulates CXCR4 expression, induces apoptosis and shows anti-tumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3352 Supplier Medchemexpress LLC Supplier No. HYN001110MG
Medchemexpress, HY-N0011 10mg Baohuoside I CAS:113558-15-9 Baohuoside I, a flavonoid isolated from Epimedium koreanum Nakai, acts as an inhibitor of CXCR4 , downregulates CXCR4 expression, induces apoptosis and shows anti-tumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.