Learn More
Enzo Life Sciences BAP1 (human), (recombinant) (His-tag) (50 µg)
SDP
Supplier:  Enzo Life Sciences BMLUW98550050
Encompass_Preferred
Alternative name: BRCA1 associated protein 1, Ubiquitin C terminal hydrolase BAP1. Purity: ≥90% (SDS-PAGE). Formulation: Liquid. In 50mM Na-phosphate, pH 8.0, 300mM NaCl. Source: Produced in E. coli. Human BAP1 is fused at the N-terminus to a His-tag. Long Term Storage: -80°C.
Catalog No. 50-201-2336