Learn More
Apexbio Technology LLC BAPTA 85233-19-8 200mg
SDP
Catalog No. 5000087685
Encompass_Preferred
BAPTA (CAS 85233-19-8) is a cell-permeant calcium chelator widely employed in biomedical research to modulate intracellular Ca concentrations By selectively binding cytosolic calcium ions BAPTA effectively buffers rapid changes in Ca levels thereby influencing calcium-dependent signaling pathways and cellular metabolic processes Its application is particularly valuable in neurobiology and cell physiology studies investigating the regulatory roles of intracellular calcium in neuronal activity signal transduction and other calcium-mediated cellular functions
Catalog No. 50-000-87685 Supplier Apexbio Technology LLC Supplier No. B7187200
BAPTA (CAS 85233-19-8) is a cell-permeant calcium chelator widely employed in biomedical research to modulate intracellular Ca concentrations By selectively binding cytosolic calcium ions BAPTA effectively buffers rapid changes in Ca levels thereby influencing calcium-dependent signaling pathways and cellular metabolic processes Its application is particularly valuable in neurobiology and cell physiology studies investigating the regulatory roles of intracellular calcium in neuronal activity signal transduction and other calcium-mediated cellular functions