Learn More
Medchemexpress LLC Bapta | 85233-19-8 | MFCD00036255 | 98.1% | 476.43 g/mol | C22H24N2O10 | 200 MG
SDP
Catalog No. 5000215216
Encompass_Preferred
Bapta is a selective calcium (Ca2+) chelator used in biochemical and cellular research to buffer intracellular calcium and probe calcium-dependent signaling. It exhibits high selectivity over magnesium and can act as a calcium indicator; it has also been reported to inhibit phospholipase C independent of its chelating activity.
  • Selective chelation of calcium with high magnesium selectivity.
  • Functions as an intracellular calcium buffer and indicator.
  • Useful for studying calcium-dependent signaling and calcium influx.
  • Can inhibit phospholipase C independent of calcium chelation.
  • Available in solid form and as ready-to-use solution formulations.
Catalog No. 50-002-15216 Supplier Medchemexpress LLC Supplier No. HY100168200MG
Bapta is a selective calcium (Ca2+) chelator used in biochemical and cellular research to buffer intracellular calcium and probe calcium-dependent signaling. It exhibits high selectivity over magnesium and can act as a calcium indicator; it has also been reported to inhibit phospholipase C independent of its chelating activity.
  • Selective chelation of calcium with high magnesium selectivity.
  • Functions as an intracellular calcium buffer and indicator.
  • Useful for studying calcium-dependent signaling and calcium influx.
  • Can inhibit phospholipase C independent of calcium chelation.
  • Available in solid form and as ready-to-use solution formulations.