Learn More
Apexbio Technology LLC BAPTA-AM, 126150-97-8, MFCD00036696, 10 mg
SDP
Supplier:  Apexbio Technology LLC B475810
Encompass_Preferred
MF: C34H40N2O18, MW: 764.68, Solubility: 16.3mg/mL in DMSO with gentle warming. BAPTA-AM is a selective calcium chelator.
Catalog No. 50-101-0334