Learn More
Apexbio Technology LLC BAPTA-AM 126150-97-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062845
Encompass_Preferred
BAPTA-AM (CAS 126150-97-8) is a selective membrane-permeable calcium (Ca ) chelator widely utilized in biomedical research After entering cells intracellular esterases cleave BAPTA-AM releasing the active BAPTA which binds free Ca to modulate calcium-dependent cellular processes In human leukemia HL-60 and U937 cells BAPTA-AM induces apoptosis characterized by DNA fragmentation and reduced c-jun expression Additionally it inhibits voltage-gated and Ca -activated potassium currents in bovine chromaffin cells and blocks hERG Kv1 3 and Kv1 5 channels in HEK293 cells (IC 1 2 1 5 M) In mice models BAPTA-AM alters ethanol-induced behavioral responses and consumption highlighting its applicability in neuroscience and apoptosis research
Catalog No. 50-000-62845 Supplier Apexbio Technology LLC Supplier No. B47585.1
 Restricted Item
Add to Cart
Add to Cart
BAPTA-AM (CAS 126150-97-8) is a selective membrane-permeable calcium (Ca ) chelator widely utilized in biomedical research After entering cells intracellular esterases cleave BAPTA-AM releasing the active BAPTA which binds free Ca to modulate calcium-dependent cellular processes In human leukemia HL-60 and U937 cells BAPTA-AM induces apoptosis characterized by DNA fragmentation and reduced c-jun expression Additionally it inhibits voltage-gated and Ca -activated potassium currents in bovine chromaffin cells and blocks hERG Kv1 3 and Kv1 5 channels in HEK293 cells (IC 1 2 1 5 M) In mice models BAPTA-AM alters ethanol-induced behavioral responses and consumption highlighting its applicability in neuroscience and apoptosis research