Learn More
Aat Bioquest BAPTA AM UltraPure Grade CAS 126150-97-8 (25 mg)
SDP
Catalog No. 502249899
Encompass_Preferred
BAPTA AM is cell-permeable version of BAPTA 12-biso-aminophenoxyethane-NNNN-tetraacetic acid a cell-permeable calcium chelator. This catalog is for 25 mg.
This item is not returnable. View return policy
Catalog No. 50-224-9899 Supplier Aat Bioquest Supplier No. 21002
BAPTA AM is cell-permeable version of BAPTA 12-biso-aminophenoxyethane-NNNN-tetraacetic acid a cell-permeable calcium chelator. This catalog is for 25 mg.