Learn More
Abcam BAPTA-AM, Ca2+ chelator, 25MG
Supplier:  Abcam AB12050325MG
Encompass_Preferred
MW 764.7 Da, Purity >97%. Selective Ca2+ chelator. Cell-permeable analog of BAPTA (ab120449). Useful for manipulation of intracellular free Ca2+ levels. Shows varied biological activity. Blocks hKv1.5, Kv11.1 (hERG) and hKv1.3 channels (Ki values are 1.23, 1.30 and 1.45 μM, respectively).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1079689