Learn More
Cayman Chemical BAPTA AM, 126150-97-8, 25 mg
SDP
Supplier:  Cayman Chemical 1555125
Encompass_Preferred
Molecular Formula C34H40N2O18, Purity ≥98%, Formula Weight 764.7, Synonyms: N,N'-[1,2-ethanediylbis(oxy-2,1-phenylene)]bis[N-[2-[(acetyloxy)methoxy]-2-oxoethyl]-1,1'-bis[(acetyloxy)methyl] ester-glycine, BAPTA Acetoxymethyl ester; A cell permeable analog of BAPTA that binds calcium only after the AM group is removed by cytoplasmic esterases; commonly used at 10-100 M to evaluate the role of intracellular calcium in cell signaling; also inhibits Kv channels, including Kv1.3, Kv1.5, and Kv11.1 (Ki = 1.45, 1.23, and 1.30 M, respectively).
Catalog No. NC1321290