Learn More
Enzo Life Sciences BAPTA-AM (25mg). CAS: 139890-68-9
SDP
Supplier:  Enzo Life Sciences BMLCA4110025
Encompass_Preferred
BAPTA is a Ca2+ chelator with 105-fold greater affinity for Ca2+ than for Mg2+. BAPTA-AM is a cell-permeable derivative which is widely used as an intracellular calcium sponge. Induces inactivation of protein kinase C. Also inhibits thapsigargin-induced apoptosis in rat thymocytes. Alternative name: 1,2-bis-(o-Aminophenoxy)-ethane-N,N,N',N'-tetraacetic acid, tetraacetoxymethyl ester. Purity: ≥90% (HPLC). Solubility: Soluble in DMSO (25mg/ml), dimethyl formamide (25mg/ml) or 100% ethanol (5mg/ml, warm). Long Term Storage: -20°C.
Catalog No. 50-201-0390