Learn More
AdipoGen BAPTA tetraethyl ester (10 g)
SDP
Supplier:  AdipoGen CDXB0287G010
Encompass_Preferred
Chemical. CAS: 73630-07-6. Formula: C30H40N2O10. MW: 588.6. Building block used for synthesis of calcium indicators and calcium chelating agents.
Catalog No. 50-205-4309