Learn More
Medchemexpress LLC Bapta tetrapotassium | 73630-08-7 | MFCD00070080 | 99.5% | 628.79 | C22H20K4N2O10 | 250 MG
SDP
Catalog No. 5000215274
Encompass_Preferred
BAPTA tetrapotassium is the tetrapotassium salt of BAPTA, a selective calcium chelator commonly used to buffer intracellular Ca2+ and study calcium-dependent signaling. It is provided as a high-purity solid for laboratory applications and is compatible with aqueous buffer systems.
Catalog No. 50-002-15274 Supplier Medchemexpress LLC Supplier No. HY100168B250MG
BAPTA tetrapotassium is the tetrapotassium salt of BAPTA, a selective calcium chelator commonly used to buffer intracellular Ca2+ and study calcium-dependent signaling. It is provided as a high-purity solid for laboratory applications and is compatible with aqueous buffer systems.